Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
The function of Mhc is muscle contraction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha cardiac MHC; alpha myosin; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; epididymis luminal protein 71; MyHC-2x; MyHC-alpha; MyHC-IIx/d; Myosin heavy chain 1; Myosin heavy chain 2x; Myosin heavy chain 6; Myosin heavy chain IIx/d; myosin heavy chain polypeptide 6 cardiac muscle adult; Myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; Myosin heavy chain, skeletal muscle, adult 1; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); Myosin-1; Myosin-6; type 2X myosin heavy chain
Gene Aliases: A530084A17Rik; A830009F23Rik; AA517445; alpha-MHC; ASD3; CMD1EE; CMH14; HEL71; IId; IId/x; MdMs; MHC-2X/D; MHC2X/D; MYH1; MYH6; MYHa; MYHC; MyHC-2x; MyHC-2X/D; Myhc-a; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; MYHCA; Myhs-f; Myhs-f2; MYHSA1; Myhsf2; SSS3
UniProt ID: (Human) P12882, (Human) P13533, (Mouse) Q5SX40, (Mouse) Q02566
Entrez Gene ID: (Human) 4619, (Human) 4624, (Mouse) 17879, (Rat) 287408, (Mouse) 17888, (Rat) 29556
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support