Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Myosin is a protein which is known to interact with actin in muscle and non-muscle cells. It contains two identical heavy chains and four light chains. Myosin molecules consist of two major regions: tails (rods) and heads. They aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. They are designated at type I (slow twitch) and type II (fast twitch).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis luminal protein 71; MyHC-2x; MyHC-IIx/d; Myosin heavy chain 1; myosin heavy chain 2d; Myosin heavy chain 2x; Myosin heavy chain IIx/d; Myosin heavy chain, skeletal muscle, adult 1; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy polypeptide 1, skeletal muscle, adult; Myosin-1; type 2X myosin heavy chain
Gene Aliases: A530084A17Rik; HEL71; IId; IId/x; MdMs; MHC-2X/D; MHC2X/D; MYH1; MYHa; MYHC; MYHC-2d; MYHC-2X; MyHC-2X/D; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; Myhs-f; Myhs-f2; MYHSA1; Myhsf2
UniProt ID: (Horse) Q8MJV0, (Human) P12882, (Mouse) Q5SX40
Entrez Gene ID: (Horse) 791235, (Human) 4619, (Mouse) 17879, (Rat) 287408
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support