Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The transcript produced from this gene is bi-cistronic and can encode both myotrophin and leucine zipper protein 6. The myotrophin protein is associated with cardiac hypertrophy, where it is involved in the conversion of NFkappa B p50-p65 heterodimers to p50-p50 and p65-p65 homodimers. This protein also has a potential function in cerebellar morphogenesis, and it may be involved in the differentiation of cerebellar neurons, particularly of granule cells. A cryptic ORF at the 3' end of this transcript uses a novel internal ribosome entry site and a non-AUG translation initiation codon to produce leucine zipper protein 6, a 6. 4 kDa tumor antigen that is associated with myeloproliferative disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Granule cell differentiation protein; growth factor; Myotrophin; Protein V-1; stimulates myocyte growth; V-1 protein
Gene Aliases: 5033418D15Rik; GCDP; MTPN; V-1; V1
UniProt ID: (Human) P58546, (Rat) P62775, (Mouse) P62774
Entrez Gene ID: (Human) 136319, (Rat) 79215, (Mouse) 14489
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support