Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody is predicted to react with porcine based on sequence homology.
NRAS (GTPase NRas) is a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. This shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. This protein, which has intrinsic GTPase activity, is activated to a GTP-bound form by a GTPase activating protein and inactivated to a GDP-bound form by a guanine nucleotide-exchange factor. Defects in the gene encoding this protein are a cause of juvenile myelomonocytic leukemia (JMML) and Noonan syndrome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTPase NRas; N-ras protein part 4; Neuroblastoma RAS; neuroblastoma RAS viral (v-ras) oncogene homolog; RAS; RASN; Transforming protein N-Ras; v-ras neuroblastoma RAS viral oncogene homolog
Gene Aliases: ALPS4; CMNS; HRAS1; N-ras; NCMS; NRAS; NRAS1; NS6
UniProt ID: (Human) P01111
Entrez Gene ID: (Human) 4893
Molecular Function: small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support