Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
In Western blots, approximately 20% cross-reactivity with recombinant human (rh) ULBP-2 is observed and no cross-reactivity with rhULBP-1 or rhULBP-3 is observed.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
This gene belong to the RAET1 family, which consists of major histocompatibility complex (MHC) class I-related genes located in a cluster on chromosome 6q24.2-q25.3. This and RAET1G protein differ from other RAET1 proteins in that they have type I membrane-spanning sequences at their C termini rather than glycosylphosphatidylinositol anchor sequences. This protein functions as a ligand for NKG2D receptor, which is expressed on the surface of several types of immune cells, and is involved in innate and adaptive immune responses. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Aug 2011].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lymphocyte effector toxicity activation ligand; MGC125309; N2DL-4; NKG2D ligand 4; RAE-1-like transcript 4; RAE1 like transcript 4; Retinoic acid early transcript 1E; UL16 binding protein 4; UL16-binding protein 4; ULBP 4; ULBP4MGC125308
Gene Aliases: bA350J20.7; LETAL; N2DL-4; N2DL4; NKG2DL4; RAET1E; RAET1E2; RL-4; ULBP4; UNQ1867/PRO4303
UniProt ID: (Human) Q8TD07
Entrez Gene ID: (Human) 135250
Molecular Function: major histocompatibility complex protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support