Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAARAVLDEF TAPAEKAELL EQSRGRIEGL FGVSLAVLGA LGAEEPLPAR IWLQLCGAQE AVHSAKEYIK GICEPELEER ECYPKDMHCI FVGAESLFLK SLIQDTCADL CILDIGLLGI RGSAEAVVMA RSHIQQFVKL FENKENLPSS QKESEVKREF KQFVEAHADN YTMDLLILPT SLKKELLTLT QGEENLFETG DDEVIEMRDS QQTEFTQNAA TGLNISRDET VLQEEARNKA GTPVSELTKQ MDTVLSSSPD VLFDPINGLT PDEEALSNER ICQKRRFSDS EERHTKKQFS LENVQEGEIL HDAKTLAGNV IADLSDSSAD SENLSPDIKE TTEEMEYNIL VNFFKTMGYS QEIVEKVIKV YGPSTEPLLL LEEIEKENKR FQEDREFSAG TVYPETNKTK NKGVYSSTNE LTTDSTPKKT QAHTQQNMVE KFSQLPFKVE AKPCTSNCRI NTFRTVPIEQ KHEVWGSNQN YICNTDPETD GLSPSVASPS PKEVNFVSRG ASSHQPRVPL FPENGLHQQP EPLLPNNMKS ACEKRLGCCS SPHSKPNCST LSPPMPLPQL LPSVTDARSA GPSDHIDSSV TGVQRFRDTL KIPYKLELKN EPGRTDLKHI VIDGSNVAIT HGLKKFFSCR GIAIAVEYFW KLGNRNITVF VPQWRTRRDP NVTEQHFLTQ LQELGILSLT PARMVFGERI ASHDDRFLLH LADKTGGIIV TNDNFREFVN ESVSWREIIT KRLLQYTFVG DIFMVPDDPL GRSGPRLEEF LQKEVCLRDM QPLLSALPNV GMFDPSFRVP GTQAASTSHQ PPTRIQGAPS SHWLPQQPHF PLLPALPSLQ QNLPMPAQRS SAETNELREA LLKIFPDSEQ RLKIDQILVA HPYMKDLNAL SAMVLD
Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N4BP1; NEDD4-binding protein 1
Gene Aliases: KIAA0615; N4BP1
UniProt ID: (Human) O75113
Entrez Gene ID: (Human) 9683
Molecular Function: endoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support