Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is Hela cell lysate.
PA5-21099 can be used with blocking peptide PEP-1213.
Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N4BP1; Nedd4 binding protein 1; NEDD4-binding protein 1
Gene Aliases: AI481586; C81621; KIAA0615; N4BP1; RGD1305179
UniProt ID: (Human) O75113, (Mouse) Q6A037
Entrez Gene ID: (Human) 9683, (Rat) 291921, (Mouse) 80750
Molecular Function: endoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support