Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Single-stranded DNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSSB2; Nucleic acid-binding protein 1; oligonucleotide/oligosaccharide-binding fold containing 2A; Oligonucleotide/oligosaccharide-binding fold-containing protein 2A; Sensor of single-strand DNA complex subunit B2; Sensor of ssDNA subunit B2; Single-stranded DNA-binding protein 2; SOSS complex subunit B2; SOSS-B2
Gene Aliases: NABP1; OBFC2A; SOSS-B2; SSB2
UniProt ID: (Human) Q96AH0
Entrez Gene ID: (Human) 64859
Molecular Function: nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support