Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Single-stranded DNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSSB2; Nucleic acid-binding protein 1; oligonucleotide/oligosaccharide-binding fold containing 2A; Oligonucleotide/oligosaccharide-binding fold-containing protein 2A; Sensor of single-strand DNA complex subunit B2; Sensor of ssDNA subunit B2; Single-stranded DNA-binding protein 2; SOSS complex subunit B2; SOSS-B2
Gene Aliases: 4930434H03Rik; 4930442A21Rik; 4930488J04Rik; 4933440J18Rik; 5830411E10Rik; AI852561; NABP1; Nbp1; OBFC2A; RGD1306658; SOSS-B2; SSB2
UniProt ID: (Human) Q96AH0, (Rat) Q5FVP2, (Mouse) Q8BGW5
Entrez Gene ID: (Human) 64859, (Dog) 478847, (Rat) 363227, (Mouse) 109019
Molecular Function: nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support