Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDMKPLVVL GLPAPTAPSG CLSFWEAKAQ LAKSCKVLVD ALRHNAAAAV PFFGGGSVLR AAEPAPHASY GGIVSVETDL LQWCLESGSI PILCPIGETA ARRSVLLDSL EVTASLAKAL RPTKIIFLNN TGGLRDSSHK VLSNVNLPAD LDLVCNAEWV STKERQQMRL IVDVLSRLPH HSSAVITAAS TLLTELFSNK GSGTLFKNAE RMLRVRSLDK LDQGRLVDLV NASFGKKLRD DYLASLRPRL HSIYVSEGYN AAAILTMEPV LGGTPYLDKF VVSSSRQGQG SGQMLWECLR RDLQTLFWRS RVTNPINPWY FKHSDGSFSN KQWIFFWFGL ADIRDSYELV NHAKGLPDSF HKPASDPGS (167-534 aa encoded by BC111713)
NAGS is a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia.The N-acetylglutamate synthase gene encodes a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. This gene may regulate ureagenesis by altering NAG availability and, thereby, CPSI activity. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amino-acid acetyltransferase; amino-acid N-acetyltransferase; MGC133025; N-acetylglutamate synthase, mitochondrial
Gene Aliases: 1700120E20Rik; AGAS; ARGA; NAGS; RGD1565783
UniProt ID: (Human) Q8N159, (Mouse) Q8R4H7
Entrez Gene ID: (Human) 162417, (Rat) 303563, (Mouse) 217214
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support