Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MVSSAQMGFN LQALLEQLSQ DELSKFKYLI TTFSLAHELQ KIPHKEVDKA DGKQLVEVLT THCDSYWVEM ASLQVFEKMH RMDLSERAKD EVREAALKSF NKRKPLSLGI TRKERPPLDV DEMLERFKTE AQAFTETKGN VICLGKEVFK GKKPDKDNRC RYILKTKFRE MWKSWPGDSK EVQVMAERYK MLIPFSNPRV LPGPFSYTVV LYGPAGLGKT TLAQKLMLDW AEDNLIHKFK YAFYLSCREL SRLGPCSFAE LVFRDWPELQ DDIPHILAQA RKILFVIDGF DELGAAPGAL IEDICGDWEK KKPVPVLLGS LLNRVMLPKA ALLVTTRPRA LRDLRILAEE PIYIRVEGFL EEDRRAYFLR HFGDEDQAMR AFELMRSNAA LFQLGSAPAV CWIVCTTLKL QMEKGEDPVP TCLTRTGLFL RFLCSRFPQG AQLRGALRTL SLLAAQGLWA QTSVLHREDL ERLGVQESDL RLFLDGDILR QDRVSKGCYS FIHLSFQQFL TALFYTLEKE EEEDRDGHTW DIGDVQKLLS GVERLRNPDL IQAGYYSFGL ANEKRAKELE ATFGCRMSPD IKQELLRCDI SCKGGHSTVT DLQELLGCLY ESQEEELVKE VMAQFKEISL HLNAVDVVPS SFCVKHCRNL QKMSLQVIKE NLPENVTASE SDAEVERSQD DQHMLPFWTD LCSIFGSNKD LMGLAINDSF LSASLVRILC EQIASDTCHL QRVVFKNISP ADAHRNLCLA LRGHKTVTYL TLQGNDQDDM FPALCEVLRH PECNLRYLGL VSCSATTQQW ADLSLALEVN QSLTCVNLSD NELLDEGAKL LYTTLRHPKC FLQRLS
NALP2 belongs to a family of cytoplasmic proteins that have been implicated in cell responses to apoptotic and inflammatory stimuli. Unlike the prototypical NALP protein NALP1, NALP2 only contains a NACHT domain, leucine rich repeat (LRR), and pyrin-containing domain (PYD). This protein interacts with the adapter protein ASC in addition to CARD8 and caspase-1 to form an inflammasome with high proIL-1-b-processing activity and is thought to function as a modulator of NF-kappa-B and procaspase-1 activation in macrophages. It has also been suggested that NALP2, in addition to other NALP family members, can act as innate sensors of pathogens in a manner similar to the toll-like receptors (TLRs). At least two alternatively spliced transcript variants encoding distinct isoforms have been found for NALP2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20510; NACHT, leucine rich repeat and PYD containing 2; NACHT, LRR and PYD containing protein 2; NACHT, LRR and PYD domains-containing protein 2; NLR family, pyrin domain containing 2; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 2; Nucleotide-binding site protein 1; PYRIN domain and NACHT domain-containing protein 1; PYRIN-containing APAF1-like protein 2
Gene Aliases: CLR19.9; NALP2; NBS1; NLRP2; PAN1; PYPAF2
UniProt ID: (Human) Q9NX02
Entrez Gene ID: (Human) 55655
Molecular Function: non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support