Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGTFDLWTD YLGLAHLVRA LSGKEGPETR LSPQPEPEPM LEPVSALEPM PAPESVPVPG PKDQKRSLES SPAPERLCSF CKHNGESRAI YQSHVLKDEA GRVLCPILRD YVCPQCGATR ERAHTRRFCP LTGQGYTSVY SHTTRNSAGK KLVRPDKAKT QDTGHRRGGG GGAGFRGAGK SEPSPSCSPS MST (1-192 aa encoded by BC101209)
Nanos is a zinc-finger containing, RNA-binding protein that has been implicated in germ cell development in both invertebrates and vertebrates. In Drosophila, Nanos represses apoptosis during development to ensure proper germ-line development. Unlike Nanos1 whose expression in mice is dispensable, the Nanos2 and Nanos3 proteins are required for germ cell development. Nanos2-null primordial germ cells (PGCs) die only in the male gonads and show no defects in females, while Nanos3-null PGCs are lost during the migration stage regardless of sex. Nanos2 and Nanos3 have distinct expression patterns during embryo development, suggesting that these two proteins do not have redundant functions. However, expression of Nanos2 can at least partially replace Nanos3 function in a Nanos3-null background. Nanos3 expression can not rescue Nanos2-null defects.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nanos homolog 3; NOS-3
Gene Aliases: NANOS1L; NANOS3; NOS3; ZC2HC12C
UniProt ID: (Human) P60323, (Mouse) P60324
Entrez Gene ID: (Human) 342977, (Mouse) 244551, (Rat) 288909
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support