Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDNAGKERE AVQLMAEAEK RVKASHSFLR GLFGGNTRIE EACEMYTRAA NMFKMAKNWS AAGNAFCQAA KLHMQLQSKH DSATSFVDAG NAYKKADPQE AINCLNAAID IYTDMGRFTI AAKHHITIAE IYETELVDIE KAIAHYEQSA DYYKGEESNS SANKCLLKVA AYAAQLEQYQ KAIEIYEQVG ANTMDNPLLK YSAKDYFFKA ALCHFIVDEL NAKLALEKYE EMFPAFTDSR ECKLLKKLLE AHEEQNSEAY TEAVKEFDSI SRLDQWLTTM LLRIKKSIQG DGEGDGDLK (1-298 aa encoded by BC060840 )
Required for vesicular transport between the endoplasmic reticulum and the Golgi apparatus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta soluble NSF attachment protein; Beta-soluble NSF attachment protein; beta-soluble NSF attachment protein {ECO:0000250|UniProtKB:P28663}; brain protein 14; Brain protein I47; N-ethylmaleimide sensitive fusion protein attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta; N-ethylmaleimide-sensitive factor attachment protein beta {ECO:0000250|UniProtKB:P28663}; N-ethylmaleimide-sensitive factor attachment protein, beta; napb {ECO:0000250|UniProtKB:P28663}; SNAP-beta; SNAP-beta {ECO:0000250|UniProtKB:P28663}
Gene Aliases: b-SNAP; Brp14; E161; I47; NAPB; SNAP-BETA; SNAPB; SNARE
UniProt ID: (Human) Q9H115, (Mouse) P28663
Entrez Gene ID: (Human) 63908, (Rat) 499903, (Mouse) 17957
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support