Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: TAAMVMPIA EAVVQQIINA EAEVEATQMT YFNGSTNHGL EIDESVNGHE INERKEKTKP VPGYNNDTGK ISSKVELEKN SGMRTKYRTK KGHVTRKLTC LCIAYSST (143-249 aa encoded by BC148686)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)/sulfate cotransporter; NaSi-1; Renal sodium/sulfate cotransporter; solute carrier family 13 (sodium/sulfate symporters), member 1; solute carrier family 13 (sodium/sulphate symporters), member 1; Solute carrier family 13 member 1
Gene Aliases: AI314066; AI987826; NAS1; NaSi-1; NASI1; SLC13A1
UniProt ID: (Human) Q9BZW2, (Mouse) Q9JHI4
Entrez Gene ID: (Human) 6561, (Mouse) 55961
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support