Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKGSRIELG DVTPHNIKQL KRLNQVIFPV SYNDKFYKDV LEVGELAKLA YFNDIAVGAV CCRVDHSQNQ KRLYIMTLGC LAPYRRLGIG TKMLNHVLNI CEKDGTFDNI YLHVQISNES AIDFYRKFGF EIIETKKNYY KRIEPADAHV LQKNLKVPSG QNADVQKTDN (1-169 aa encoded by BC012731)
NAT13 is a probable catalytic component of the ARD1A-NARG1 complex which displays alpha (N-terminal) acetyltransferase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hNaa50p; hNAT5; hSAN; Mak3 homolog; Mak3p homolog; N-acetyltransferase 13; N-acetyltransferase 13 (GCN5-related); N-acetyltransferase 5; N-acetyltransferase NAT13; N-acetyltransferase san homolog; N-alpha-acetyltransferase 50; N-alpha-acetyltransferase 50, NatE catalytic subunit; N-epsilon-acetyltransferase 50; Nalpha acetyltransferase 50; NatE catalytic subunit
Gene Aliases: 2600005K24Rik; 2810441M03Rik; AW112078; MAK3; Mak3p; NAA50; NAT13; NAT13P; NAT5; NAT5P; SAN
UniProt ID: (Human) Q9GZZ1, (Mouse) Q6PGB6
Entrez Gene ID: (Human) 80218, (Rat) 288108, (Mouse) 72117
Molecular Function: acetyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support