Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTTLRAFTC DDLFRFNNIN LDPLTETYGI PFYLQYLAHW PEYFIVAEAP GGELMGYIMG KAEGSVAREE WHGHVTALSV APEFRRLGLA AKLMELLEEI SERKGGFFVD LFVRVSNQVA VNMYKQLGYS VYRTVIEYYS ASNGEPDEDA YDMRKALSRD TEKKSIIPLP HPVRPEDIE (1-178 aa encoded by BC005181 )
Catalytic subunit of the NatB complex which catalyzes acetylation of the N-terminal methionine residues of peptides beginning with Met-Asp, Met-Glu, Met-Asn and Met-Gln. Proteins with cell cycle functions are overrepresented in the pool of NatB substrates. Required for maintaining the structure and function of actomyosin fibers and for proper cellular migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Methionine N-acetyltransferase; N-acetyltransferase 3 homolog; N-acetyltransferase 5; N-acetyltransferase 5 (ARD1 homolog, S. cerevisiae); N-acetyltransferase 5 (GCN5-related, putative); N-acetyltransferase 5, ARD1 subunit (arrest-defective 1, S. cerevisiae, homolog); N-alpha-acetyltransferase 20; N-alpha-acetyltransferase 20, NatB catalytic subunit; N-terminal acetyltransferase B complex catalytic subunit NAA20; N-terminal acetyltransferase B complex catalytic subunit NAT5; N-terminal acetyltransferase complex ARD1 subunit; NatB catalytic subunit; NatB complex subunit NAT5
Gene Aliases: 1500004D14Rik; 2900026I01Rik; AU041458; D2Ertd186e; dJ1002M8.1; NAA20; NAT3; NAT3P; NAT5; NAT5P
UniProt ID: (Human) P61599, (Mouse) P61600
Entrez Gene ID: (Human) 51126, (Mouse) 67877, (Rat) 362228
Molecular Function: acetyltransferase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support