Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: TRYVDIALRT DMSDITKSYL SECGSCFWVG ESEEKVVGTV GALPVDDPTL REKRLQLFHL SVDNEHRGQG IAKALVRTVL QFARDQGYSE VVLDTSN
The protein encoded by this gene is highly similar to the N-acetyltransferase 8 (NAT8) gene product, which is a kidney and liver protein with homology to bacterial acetyltransferases involved in drug resistance. This gene is localized on chromosome 2 in the vicinity of the NAT8 gene and may represent a pseudogene of NAT8. This gene contains two polymorphic nonsense mutations that disrupt the active site of the protein. The full-length product of this gene contains a complete acetyltransferase domain and is identical in length to NAT8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetyltransferase 1; ATase1; camello-like protein 2; N-acetyltransferase 8B (GCN5-related, putative, gene/pseudogene); N-acetyltransferase Camello 2; probable N-acetyltransferase 8B
Gene Aliases: CML2; Hcml2; NAT8BP
Entrez Gene ID: (Human) 51471
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support