Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: NAV2.
The vitamin A metabolite, all-trans retinoic acid (atRA), plays an important role in neuronal development, including neurite outgrowth. RAINB1 is an atRA-responsive gene. [supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Helicase APC down-regulated 1; helicase, APC down-regulated 1; Neuron navigator 2; Pore membrane and/or filament-interacting-like protein 2; retinoic acid inducible gene in neuroblastoma 1; Retinoic acid inducible in neuroblastoma 1; Steerin-2; Unc-53 homolog 2; unc53H2
Gene Aliases: HELAD1; KIAA1419; NAV2; POMFIL2; RAINB1; STEERIN2; UNC53H2
UniProt ID: (Human) Q8IVL1
Entrez Gene ID: (Human) 89797
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support