Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
This antibody has specificity for Human NCAPH.
This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: barren homolog 1; Barren homolog protein 1; Chromosome-associated protein H; Condensin complex subunit 2; hCAP-H; Non-SMC condensin I complex subunit H; non-SMC condensin I complex, subunit H; XCAP-H homolog
Gene Aliases: BRRN; BRRN1; CAP-H; CAPH; KIAA0074; NCAPH
UniProt ID: (Human) Q15003
Entrez Gene ID: (Human) 23397
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support