Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSRRRHSDEN DGGQPHKRRK TSDANETEDH LESLICKVGE KSACSLESNL EGLAGVLEAD LPNYKSKILR LLCTVARLLP EKLTIYTTLV GLLNARNYNF GGEFVEAMIR QLKESLKANN YNEAVYLVRF LSDLVNCHVI AAPSMVAMFE NFVSVTQEED VPQVRRDWYV YAFLSSLPWV GKELYEKKDA EMDRIFANTE SYLKRRQKTH VPMLQVWTAD KPHPQEEYLD CLWAQIQKLK KDRWQERHIL RPYLAFDSIL CEALQHNLPP FTPPPHTEDS VYPMPRVIFR MFDYTDDPEG PVMPGSHSVE RFVIEENLHC IIKSHWKERK TCAAQLVSYP GKNKIPLNYH IVEVIFAELF QLPAPPHIDV MYTTLLIELC KLQPGSLPQV LAQATEMLYM RLDTMNTTCV DRFINWFSHH LSNFQFRWSW EDWSDCLSQD PESPKPKFVR EVLEKCMRLS YHQRILDIVP PTFSALCPAN PTCIYKYGDE SSNSLPGHSV ALCLAVAFKS KATNDEIFSI LKDVPNPNQD DDDDEGFSFN PLKIEVFVQT LLHLAAKSFS HSFSALAKFH EVFKTLAESD EGKLHVLRVM FEVWRNHPQM IAVLVDKMIR TQIVDCAAVA NWIFSSELSR DFTRLFVWEI LHSTIRKMNK HVLKIQKELE EAKEKLARQH KRRSDDDDRS SDRKDGVLEE QIERLQEKVE SAQSEQKNLF LVIFQRFIMI LTEHLVRCET DGTSVLTPWY KNCIERLQQI FLQHHQIIQQ YMVTLENLLF TAELDPHILA VFQQFCALQA
May be involved in protein trafficking from the plasma membrane to the early endosome (EE) as well as in homotypic fusion of endocytic organelles. Mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80 kDa nuclear cap-binding protein; CBP80; MGC2087; NCBP 80 kDa subunit; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; Nuclear cap-binding protein subunit 1; RP11-546O6.1
Gene Aliases: CBP80; NCBP; NCBP1; Sto1
UniProt ID: (Human) B2R718
Entrez Gene ID: (Human) 4686
Molecular Function:
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support