Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
NCKAP5 (NCK Associated Protein 5) is a Protein Coding gene. An important paralog of this gene isNCKAP5L.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NAP-5; NCK-associated protein 5; Peripheral clock protein; peripheral clock protein 2
Gene Aliases: 8430408F21; D130011D22Rik; E030049G20Rik; ERIH; ERIH1; ERIH2; Gm1548; NAP5; NCKAP5; RGD1564507
UniProt ID: (Human) O14513
Entrez Gene ID: (Human) 344148, (Rat) 363974, (Mouse) 210356
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support