Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Ca2+ plays a critical role in intracellular signaling and thus regulation of calcium balance is critical for the maintenance of this signaling function. The plasma membrane Na+/Ca2+ (NaCX) exchangers mediate Na+-dependent Ca2+ efflux in a wide variety of cell types. NaCX can move Ca2+ either into or out of cells, depending on the net Na+, Ca2+, and K+ gradient across the membrane. In mammals, at least 5 distinct genes code for the exchangers: Three NaCX (NaCX-1, NaCX-2, and NaCX-3), and two in the NaCKX family (NaCKX-1 and NaCKX-2). NaCX-1 is most prominently expressed in the heart where it plays a major role in excitation-contraction coupling (Matsuoka et al., 1993). NACX-1 may also be a target for protection of the heart and brain from ischemia and reperfusion injury (Hagihara et al., 2005; Bano et al., 2005; Choi, 2005).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)/Ca(2+)-exchange protein 1; Na+/Ca++ exchanger; Na+/Ca2+ exchanger; Na-Ca exchanger; Na/Ca exchanger; Sodium/calcium exchanger 1; sodium/calcium exchanger isoform NaCa7; solute carrier family 8 (sodium/calcium exchanger), member 1; Solute carrier family 8 member 1; solute carrier family 8, member 1
Gene Aliases: AI852629; AV344025; CNC; D930008O12Rik; Ncx; NCX1; SLC8A1
UniProt ID: (Human) P32418, (Mouse) P70414, (Rat) Q01728
Entrez Gene ID: (Human) 6546, (Mouse) 20541, (Rat) 29715
Molecular Function: secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support