Product References
CRISPR/Cas9 genome editing reveals an essential role for basigin in maintaining a nonkeratinized squamous epithelium in cornea.
FASEB bioAdvances
Woodward AM,Feeley MN,Rinaldi J,Argüeso P
Published figure using NCoR2 polyclonal antibody (Product # PA1-843) in Western Blot
Mon Nov 01 00:00:00 EDT 2021
TBL1XR1 Mutations Drive Extranodal Lymphoma by Inducing a Pro-tumorigenic Memory Fate.
Cell
Venturutti L,Teater M,Zhai A,Chadburn A,Babiker L,Kim D,Béguelin W,Lee TC,Kim Y,Chin CR,Yewdell WT,Raught B,Phillip JM,Jiang Y,Staudt LM,Green MR,Chaudhuri J,Elemento O,Farinha P,Weng AP,Nissen MD,Steidl C,Morin RD,Scott DW,Privé GG,Melnick AM
PA1-843 was used in Immunoprecipitation to show that TBL1XR1 mutations skew the humoral immune response toward generating abnormal immature memory B cells (MB), while impairing plasma cell differentiation.
Thu Jul 23 00:00:00 EDT 2020
ARNT2 Tunes Activity-Dependent Gene Expression through NCoR2-Mediated Repression and NPAS4-Mediated Activation.
Neuron
Sharma N,Pollina EA,Nagy MA,Yap EL,DiBiase FA,Hrvatin S,Hu L,Lin C,Greenberg ME
PA1-843 was used in Western Blotting to conclude that the interplay of bHLH-PAS complexes at activity-dependent regulatory elements maintains temporal control of activity-dependent gene expression and scales somatic inhibition with circuit activity.
Wed Apr 17 00:00:00 EDT 2019
Autoregulatory loop of nuclear corepressor 1 expression controls invasion, tumor growth, and metastasis.
Proceedings of the National Academy of Sciences of the United States of America
Martínez-Iglesias OA,Alonso-Merino E,Gómez-Rey S,Velasco-Martín JP,Martín Orozco R,Luengo E,García Martín R,Ibáñez de Cáceres I,Fernández AF,Fraga MF,González-Peramato P,Varona C,Palacios J,Regadera J,Aranda A
PA1-843 was used in western blot to study how nuclear corepressor 1 contributes to cancer
Tue Jan 19 00:00:00 EST 2016
PHD2/3-dependent hydroxylation tunes cardiac response to β-adrenergic stress via phospholamban.
The Journal of clinical investigation
Xie L,Pi X,Townley-Tilson WH,Li N,Wehrens XH,Entman ML,Taffet GE,Mishra A,Peng J,Schisler JC,Meissner G,Patterson C
PA1-843 was used in immunoprecipitation to investigate how PHD2 and PHD3 modulate contractile function.
Wed Jul 01 00:00:00 EDT 2015
PHD2/3-dependent hydroxylation tunes cardiac response to β-adrenergic stress via phospholamban.
The Journal of clinical investigation
Xie L,Pi X,Townley-Tilson WH,Li N,Wehrens XH,Entman ML,Taffet GE,Mishra A,Peng J,Schisler JC,Meissner G,Patterson C
PA1-843 was used in immunoprecipitation to investigate how PHD2 and PHD3 modulate contractile function.
Wed Jul 01 00:00:00 EDT 2015
NCoR1 and SMRT play unique roles in thyroid hormone action in vivo.
Molecular and cellular biology
Shimizu H,Astapova I,Ye F,Bilban M,Cohen RN,Hollenberg AN
PA1-843 was used in western blot to examine the role of coregulators of nuclear receptor action in thyroid hormone action
Sun Feb 01 00:00:00 EST 2015
NCoR1 and SMRT play unique roles in thyroid hormone action in vivo.
Molecular and cellular biology
Shimizu H,Astapova I,Ye F,Bilban M,Cohen RN,Hollenberg AN
PA1-843 was used in western blot to examine the role of coregulators of nuclear receptor action in thyroid hormone action
Sun Feb 01 00:00:00 EST 2015
Thyroid hormone signaling in vivo requires a balance between coactivators and corepressors.
Molecular and cellular biology
Vella KR,Ramadoss P,Costa-E-Sousa RH,Astapova I,Ye FD,Holtz KA,Harris JC,Hollenberg AN
PA1-843 was used in western blot to study the role of the coactivator/corepressor balance in thyroid hormone signaling
Thu May 01 00:00:00 EDT 2014
Corepressors (NCoR and SMRT) as well as coactivators are recruited to positively regulated 1α,25-dihydroxyvitamin D3-responsive genes.
The Journal of steroid biochemistry and molecular biology
Meyer MB,Pike JW
PA1-843 was used in ChIP assay to study the recruitment of both co-activators and co-repressors to vitamin D3-responsive genes
Mon Jul 01 00:00:00 EDT 2013
A FOXO3-IRF7 gene regulatory circuit limits inflammatory sequelae of antiviral responses.
Nature
Litvak V,Ratushny AV,Lampano AE,Schmitz F,Huang AC,Raman A,Rust AG,Bergthaler A,Aitchison JD,Aderem A
PA1-843 was used in ChIP assay to study the role of FOXO3 and IRF7 in minimising inflammatory damage in response to viral infection
Thu Oct 18 00:00:00 EDT 2012
A downstream intergenic cluster of regulatory enhancers contributes to the induction of CYP24A1 expression by 1alpha,25-dihydroxyvitamin D3.
The Journal of biological chemistry
Meyer MB,Goetsch PD,Pike JW
Published figure using NCoR2 polyclonal antibody (Product # PA1-843) in ChIP assay
Fri May 14 00:00:00 EDT 2010
Cooperative regulation in development by SMRT and FOXP1.
Genes & development
Jepsen K,Gleiberman AS,Shi C,Simon DI,Rosenfeld MG
PA1-843 was used in chromatin immunoprecipitation to study the roles of SMRT and FOXP1 in developmental regulation.
Sat Mar 15 00:00:00 EDT 2008
Cooperative regulation in development by SMRT and FOXP1.
Genes & development
Jepsen K,Gleiberman AS,Shi C,Simon DI,Rosenfeld MG
PA1-843 was used in chromatin immunoprecipitation to study the roles of SMRT and FOXP1 in developmental regulation.
Sat Mar 15 00:00:00 EDT 2008
Ataxin 1, a SCA1 neurodegenerative disorder protein, is functionally linked to the silencing mediator of retinoid and thyroid hormone receptors.
Proceedings of the National Academy of Sciences of the United States of America
Tsai CC,Kao HY,Mitzutani A,Banayo E,Rajan H,McKeown M,Evans RM
PA1-843 was used in western blot and immunocytochemistry to demonstrate the effect of mutant Atx1 on corepressor-dependent transcriptional pathways.
Tue Mar 23 00:00:00 EST 2004