Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cebelin; family with sequence similarity 163, member A; neuroblastoma derived secretory protein; Neuroblastoma-derived secretory protein; Protein FAM163A
Gene Aliases: A230106N23Rik; C1orf76; FAM163A; NDSP; RGD1565760
UniProt ID: (Human) Q96GL9, (Mouse) Q8CAA5
Entrez Gene ID: (Human) 148753, (Rat) 498257, (Mouse) 329274
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support