Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: IGFHKTSEKS VQSFQLKGFP FSIYGNLAVK DCCINPHSPL IRVTKSSKLE KGSLPGTDWT VFQINHSAYQ PVIFAKVKTP ENLSPSISKG AFYATIIH
This gene encodes a member of the heparan sulfate/heparin GlcNAc N-deacetylase/ N-sulfotransferase family. The encoded enzyme is a type II transmembrane protein that resides in the Golgi apparatus. This monomeric bifunctional enzyme catalyzes the N-deacetylation and N-sulfation of N-acetylglucosamine residues in heparan sulfate and heparin, which are the initial chemical modifications required for the biosynthesis of the functional oligosaccharide sequences that define the specific ligand binding activities of heparan sulfate and heparin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 3; GlcNAc N-deacetylase/ N-sulfotransferase 3; Glucosaminyl N-deacetylase/N-sulfotransferase 3; hNDST-3; N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3; N-deacetylase/N-sulfotransferase 3; N-heparan sulfate sulfotransferase 3; N-HSST 3; NDST-3
Gene Aliases: HSST3; NDST3; UNQ2544/PRO4998
UniProt ID: (Human) O95803
Entrez Gene ID: (Human) 9348
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support