Search Thermo Fisher Scientific
Sequence of this protein is as follows: MWFEILPGLS VMGVCLLIPG LATAYIHRFT NGGKEKRVAH FGYHWSLMER DRRISGVDRY YVSKGLENID
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: CI-MWFE; complex I MWFE subunit; Complex I-MWFE; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1, 7.5kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 1; NADH oxidoreductase subunit MWFE; NADH-ubiquinone oxidoreductase MWFE subunit; NADH:ubiquinone oxidoreductase (complex 1); type I dehydrogenase
基因别名: CI-MWFE; MWFE; NDUFA1; ZNF183
UniProt ID: (Human) O15239
Entrez Gene ID: (Human) 4694