Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse kidney.
Predicted reactivity: Chimpanzee (99%), Bovine (82%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a component of 42 kDa complex I, the first enzyme complex in the electron transport chain of mitochondria. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. A mutation in this gene was found in an individual with Leigh syndrome.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-42kD; complex I 42kDa subunit; Complex I-42kD; FLJ21923; MGC5103; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 10, 42kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 10, mitochondrial; NADH-ubiquinone oxidoreductase 42 kDa subunit
Gene Aliases: 2900053E13Rik; CI-42k; CI-42KD; NDUFA10
UniProt ID: (Human) O95299, (Mouse) Q99LC3
Entrez Gene ID: (Human) 4705, (Mouse) 67273
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support