Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Promo Code: RACCS25 Save on advanced culture solutions Save up to 22% on optimized formulations to streamline your neural, stem and cancer cell culture. Learn more
NDUFA8 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed to be not involved in catalysis. Mammalian complex I is composed of 45 different subunits and transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-19kD; CI-PGIV; Complex I-19kD; Complex I-PGIV; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8; NADH-ubiquinone oxidoreductase 19 kDa subunit; NADH:ubiquinone oxidoreductase PGIV subunit
Gene Aliases: 0610033L03Rik; Aa2-258; AW261656; CI-19KD; CI-PGIV; NDUFA8; PGIV
UniProt ID: (Human) P51970, (Mouse) Q9DCJ5
Entrez Gene ID: (Human) 4702, (Mouse) 68375, (Rat) 296658
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support