Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PGIVELPTLE ELKVDEVKIS SAVLKAAAHH YGAQCDKPNK EFMLCRWEEK DPRRCLEEGK LVNKCALDFF RQIKRHCA
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
NDUFA8 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed to be not involved in catalysis. Mammalian complex I is composed of 45 different subunits and transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-19kD; CI-PGIV; Complex I-19kD; Complex I-PGIV; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8; NADH-ubiquinone oxidoreductase 19 kDa subunit; NADH:ubiquinone oxidoreductase PGIV subunit
Gene Aliases: CI-19KD; CI-PGIV; NDUFA8; PGIV
UniProt ID: (Human) P51970
Entrez Gene ID: (Human) 4702
Molecular Function:
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support