Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAMTFCRL LNRCGEAARS LPLGARCFGV RVSPTGEKVT HTGQVYDDKD YRRIRFVGRQ KEVNENFAID LIAEQPVSEV ETRVIACDGG GGALGHPKVY INLDKETKTG TCGYCGLQFR QHHH
This gene encodes a subunit of the NADH:ubiquinone oxidoreductase, which is the first enzyme complex in the electron transport chain of mitochondria. This complex functions in the transfer of electrons from NADH to the respiratory chain. The subunit encoded by this gene is one of seven subunits in the iron-sulfur protein fraction. Mutations in this gene cause mitochondrial complex I deficiency, a disease that causes a wide variety of clinical disorders, including neonatal disease and adult-onset neurodegenerative disorders.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-13kD-A; complex I 13kDa subunit A; complex I, mitochondrial respiratory chain, 13-kD subunit; Complex I-13kD-A; NADH dehydrogenase (ubiquinone) Fe-S protein 6, 13kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 6, mitochondrial; NADH dehydrogenase Fe-S protein 6; NADH ubiquinone oxidoreductase; NADH-ubiquinone oxidoreductase 13 kDa-A subunit; NADH:ubiquinone oxidoreductase NDUFS6 subunit
Gene Aliases: BC059730; CI-13kA; CI-13kD-A; CI13KDA; IP13; Ip13dis; Ndub13; NDUFS6; RATIp13dis
UniProt ID: (Human) O75380, (Mouse) P52503, (Rat) P52504
Entrez Gene ID: (Human) 4726, (Rabbit) 100345307, (Chicken) 420976, (Mouse) 407785, (Rat) 29478
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support