Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: PSACRDEAGL REKDYDFPPP MRQAGRPDLR PEGVYDIPPT CTKPAGKDLH VKYNCDIPGA AEPVARRHQS LSPNHPPPQL GQSVGSQNDA YDVPRGVQFL EPPAETSEKA NPQERDGVYD VPLHNPPDAK GSRDLVDGIN RLSFSSTGST RSNMSTSSTS SKESSLSASP AQDKRLFLDP DTAIERLQRL QQALEMGVSS LMALVTTDWR CYGYMERHIN EIRTAVDKVE LF
HEF-1 is a multifunctional protein involved in integrin-based signaling that affects cell motility, growth, apoptosis and oncogenic transformation. The Cas family of docking proteins have been the subject of intense research because of their role in cell motility, growth, apoptosis and oncogenic transformation. These proteins are substrates of focal adhesion kinase (FAK) and the Src family of tyrosine kinases two active targets for drug development. HEF1 protein production increases levels of mRNA transcripts that encode proteins associated with motility, cell transformation and invasiveness, including several metalloproteinases, MLCK, p160ROCK and ErbBi. HEF1 overproduction also mediates apoptosis in epithelial-derived cell lines, including MCF7 and HeLa cells. Recent clinical studies at another institution have found that overexpression of BCAR1 (p130Cas), a related protein, is associated with tamoxifen resistance. This highlights the importance of studying the role of this family of proteins in cancer prognosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cas scaffolding protein family member 2; CAS-L; cas-like docking; CASS2; Crk-associated substrate related protein Cas-L; CRK-associated substrate-related protein; dJ49G10.2; dJ761I2.1; Enhancer of filamentation 1; Enhancer of filamentation 1 p55; hEF1; NEDD-9; Neural precursor cell expressed developmentally down-regulated protein 9; p105; p130Cas-related protein; Renal carcinoma antigen NY-REN-12
Gene Aliases: CAS-L; CAS2; CASL; CASS2; HEF1; NEDD9
UniProt ID: (Human) Q14511
Entrez Gene ID: (Human) 4739
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support