Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: NEGR1.
May be involved in cell-adhesion. May function as a trans-neural growth-promoting factor in regenerative axon sprouting in the mammalian brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a kindred of IgLON; IgLON family member 4; Kilon; kilon protein; Kindred of IgLON; Neuronal growth regulator 1; Neurotractin; UNQ2433
Gene Aliases: 5330422G01Rik; DMML2433; IGLON4; Kiaa3001; KILON; NEGR1; Ntra; UNQ2433/PRO4993
UniProt ID: (Human) Q7Z3B1, (Mouse) Q80Z24, (Rat) Q9Z0J8
Entrez Gene ID: (Human) 257194, (Mouse) 320840, (Rat) 59318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support