Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total NEGR1.
May be involved in cell-adhesion. May function as a trans-neural growth-promoting factor in regenerative axon sprouting in the mammalian brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a kindred of IgLON; IgLON family member 4; Kilon; kilon protein; Kindred of IgLON; Neuronal growth regulator 1; Neurotractin; UNQ2433
Gene Aliases: 5330422G01Rik; DMML2433; IGLON4; Kiaa3001; KILON; NEGR1; Ntra; UNQ2433/PRO4993
UniProt ID: (Human) Q7Z3B1, (Mouse) Q80Z24, (Rat) Q9Z0J8
Entrez Gene ID: (Human) 257194, (Mouse) 320840, (Rat) 59318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support