Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Recommended positive controls: 293T.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NEK2 is involved in cell cycle regulation and is essential for faithful chromosome segregation. NEK2 is a gene that encodes for a serine/threonine-protein kinase. This protein plays a crucial role in the regulation of mitosis and is primarily located in the centrosome. During the G1 phase, the protein is undetectable, but its levels progressively increase throughout the S phase, reaching their peak in late G2 phase. The gene has several alternatively spliced transcript variants that encode different isoforms with distinct C-termini. NEK2 is associated with several diseases, including Retinitis Pigmentosa 67 and Retinitis Pigmentosa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HSPK 21; NEK 2; NEK 2A; Never in mitosis A-related kinase 2; NIMA (never in mitosis gene a)-related kinase 2; NimA-like protein kinase 1; NIMA-related kinase 2; NimA-related protein kinase 2; NLK 1; protein phosphatase 1, regulatory subunit 111; Serine/threonine-protein kinase Nek2
Gene Aliases: HsPK21; NEK2; NEK2A; NLK1; PPP1R111; RP67
UniProt ID: (Human) P51955
Entrez Gene ID: (Human) 4751
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support