Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase. May be involved in activation of the SAPK/JNK pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: guanine nucleotide regulatory protein (oncogene); Net1 homolog; Neuroepithelial cell-transforming gene 1 protein; neuroepithelioma transforming gene 1; p65 Net1 proto-oncogene protein; Proto-oncogene p65 Net1; Rho guanine nucleotide exchange factor (GEF) 8; Rho guanine nucleotide exchange factor 8; RP11-116G8.1; small GTP-binding protein regulator
Gene Aliases: ARHGEF8; mNET1; NET1; NET1A
UniProt ID: (Human) Q7Z628, (Mouse) Q9Z206
Entrez Gene ID: (Human) 10276, (Mouse) 56349
Molecular Function: guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support