Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ganglioside sialidase; Ganglioside sialidasedis; Membrane sialidase; N-acetyl-alpha-neuraminidase 3; neuraminidase 3 (membrane sialidase); neuraminidase 3, membrane sialidase; sialidase 3 (membrane sialidase); Sialidase 3 (Membrane sialidase) (Ganglioside sialidase) (N-acetyl-alpha-neuraminidase 3); Sialidase-3
Gene Aliases: NEU3; SIAL3
UniProt ID: (Human) Q9UQ49, (Mouse) Q9JMH7
Entrez Gene ID: (Human) 10825, (Mouse) 50877
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support