Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total NEU4.
The protein encoded by this gene belongs to a family of glycohydrolytic enzymes, which remove terminal sialic acid residues from various sialo derivatives, such as glycoproteins, glycolipids, oligosaccharides, and gangliosides. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-acetyl-alpha-neuraminidase 4; Neuraminidase 4; neuraminidase 4 (sialidase); sialidase 4; Sialidase-4
Gene Aliases: 9330166I04; LP5125; NEU4
UniProt ID: (Human) A8K056, (Mouse) Q8BZL1
Entrez Gene ID: (Human) 129807, (Mouse) 241159, (Rat) 316642
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support