Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTKSYSESG LMGEPQPQGP PSWTDECLSS QDEEHEADKK EDDLEAMNAE EDSLRNGGEE EDEDEDLEEE EEEEEEDDDQ KPKRRGPKKK KMTKARLERF KLRRMKANAR ERNRMHGLNA ALDNLRKVVP CYSKTQKLSK IETLRLAKNY IWALSEILRS GKSPDLVSFV QTLCKGLSQP TTNLVAGCLQ LNPRTFLPEQ NQDMPPHLPT ASASFPVHPY SYQSPGLPSP PYGTMDSSHV FHVKPPPHAY SAALEPFFES PLTDCTSPSF DGPLSPPLSI NGNFSFKHEP SAEFEKNYAF TMHYPAATLA GAQSHGSIFS GTAAPRCEIP IDNIMSFDSH SHHERVMSAQ LNAIFHD (1-356 aa encoded by BC009046)
NeuroD1 is a transcriptional activator that acts as a differentiation factor during neurogenesis. It has been demonstrated to bind to the insulin gene E-box. Efficient DNA binding requires dimerization with another basic helix-loop-helix (bHLH) protein. Defects in NEUROD1 are a cause of maturity onset diabetes of the young type VI (MODY6). MODY6 is a form of non-insulin-dependent diabetes mellitus characterized by an autosomal dominant mode of inheritance, onset during young adulthood and a primary defect in insulin secretion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic helix-loop-helix factor 1; basic helix-loop-helix transcription factor; beta-cell E-box transactivator 2; Beta-cell E-box transcriptional activator 2; Beta2; BHF-1; BHF1; bHLHa3; Class A basic helix-loop-helix protein 3; neuro d1; NeuroD; NeuroD1; neurogenic differentiation 1; Neurogenic differentiation factor 1; neurogenic helix-loop-helix protein NEUROD
Gene Aliases: BETA2; BHF-1; BHLHA3; MODY6; NEUROD; NEUROD1
UniProt ID: (Human) Q13562, (Rat) Q64289, (Mouse) Q60867
Entrez Gene ID: (Human) 4760, (Rat) 29458, (Mouse) 18012
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support