Save up to 31% on select cell culture essentials

Bundle and save

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • NeuroD2 Antibodies

            NeoBiotechnologies

            NEUROD2 (Neurogenic Differentiation 2) (Transcription Factor) Monoclonal Antibody (PCRP-NEUROD2-1G1)

            View all (12) NeuroD2 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 4761-MSM1-P1ABX
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            610.00
            100 µg
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite NEUROD2 (Neurogenic Differentiation 2) (Transcription Factor) Monoclonal Antibody (PCRP-NEUROD2-1G1)

            • Antibody Testing Data (4)
            Application
            NEUROD2 (Neurogenic Differentiation 2) (Transcription Factor) Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 4

            NEUROD2 (Neurogenic Differentiation 2) (Transcription Factor) Antibody (4761-MSM1-P1ABX) in ICC/IF

            Immunofluorescence analysis of PFA-fixed HeLa cells stained using NEUROD2 Mouse Monoclonal Antibody (PCRP-NEUROD2-1G1) followed by goat anti-mouse IgG-CF488 (green). CF640A phalloidin (red).
            View Product

            NEUROD2 (Neurogenic Differentiation 2) (Transcription Factor) Antibody in Immunocytochemistry (ICC/IF)
            NEUROD2 (Neurogenic Differentiation 2) (Transcription Factor) Antibody in Flow Cytometry (Flow)
            NEUROD2 (Neurogenic Differentiation 2) (Transcription Factor) Antibody in Peptide array (ARRAY)
            NEUROD2 (Neurogenic Differentiation 2) (Transcription Factor) Antibody in SDS-PAGE (SDS-PAGE)

            Product Details

            4761-MSM1-P1ABX

            Applications
            Tested Dilution
            Publications

            Immunocytochemistry (ICC/IF)

            1-2 µg/mL
            -

            Flow Cytometry (Flow)

            1-2 µg/10^6 cells
            -

            ELISA (ELISA)

            Assay-dependent
            -

            Immunoprecipitation (IP)

            1-2 µg/100-500 µg total protein
            -

            Peptide Array (Array)

            Assay-dependent
            -
            Product Specifications

            Species Reactivity

            Human

            Host/Isotype

            Mouse / IgG2b, kappa

            Class

            Monoclonal

            Type

            Antibody

            Clone

            PCRP-NEUROD2-1G1

            Immunogen

            Recombinant full-length human NEUROD2 protein
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A/G

            Storage buffer

            PBS, pH 7.4

            Contains

            no preservative

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Target Information

            Members of the myogenic determination family are basic helix-loop-helix (bHLH) proteins that can be separated into two classes, both of which work together to activate DNA transcription. Class A proteins include the ubiquitously expressed E-box binding factors, namely E2A, ITF-2 and HEB, while class B proteins, such as MyoD, myogenin and Neuro D (BETA2), are transiently expressed and exhibit a much more limited tissue distribution. Neuro D2 (neurogenic differentiation 2), also known as NDRF, NEUROD2 or bHLHa1, is a 382 amino acid nuclear protein that contains one bHLH domain and functions to induce neurogenic differentiation, playing an important role in the maintenance and determination of cell fate.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: bHLHa1; Class A basic helix-loop-helix protein 1; NDF2; NDR2; NDRF; NeuroD-related factor; NeuroD2; neurogenic basic-helix-loop-helix protein; neurogenic differentiation 2; Neurogenic differentiation factor 2

            View more View less

            Gene Aliases: BHLHA1; NDRF; NEUROD2

            View more View less

            UniProt ID: (Human) Q15784

            View more View less

            Entrez Gene ID: (Human) 4761

            View more View less

            Function(s)

            RNA polymerase II regulatory region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding transcription factor activity, sequence-specific DNA binding transcription corepressor activity protein heterodimerization activity E-box binding basic helix-loop-helix transcription factor

            Process(es)

            behavioral fear response regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter nervous system development associative learning protein ubiquitination cerebellar cortex development positive regulation of synaptic plasticity positive regulation of neuron differentiation positive regulation of transcription from RNA polymerase II promoter neuron development positive regulation of calcium-mediated signaling positive regulation of sequence-specific DNA binding transcription factor activity cellular response to electrical stimulus cellular response to calcium ion positive regulation of synapse maturation negative regulation of nucleic acid-templated transcription negative regulation of synapse maturation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-rms4r:80/100.66.77.126:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0