Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MPARLETCI SDLDCASSSG SDLSGFLTDE EDCARLQQAA SASGPPAPAR RGAPNISRAS EVPGAQDDEQ ERRRRRGRTR VRSEALLHSL RRSRRVKAND RERNRMHNLN AALDALRSVL PSFPDDTKLT KIETLRFAYN YIWALAETLR LADQGLPGGG ARERLLPPQC VPCLPGPPSP ASDAESWGSG AAAASPLSDP SSPAASEDFT YRPGDPVFSF PSLPKDLLHT TPCFIPYH (1-237 aa encoded by BC008687)
Neurogenin 1 is a basic helix-loop-helix (bHLH) proteins are transcription factors involved in determining cell type during development. NeuroG1 is a bHLH protein with dual cell-fate specification roles. It functions during neurogenesis, and it has also been shown to inhibit the differentiation of neural stem cells into astrocytes. NeuroG1 promotes neurogenesis by functioning as a transcriptional activator, yet it inhibits astrocyte differentiation by compartmentalizing the CREB-binding protein transcription complex away from astrocyte differentiation genes and by inhibiting STAT transcription factors essential for gliogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa6; Class A basic helix-loop-helix protein 6; Helix-loop-helix protein mATH-4C; mATH4C; NeuroD3; Neurogenic basic-helix-loop-helix protein; neurogenic differentiation 3; Neurogenic differentiation factor 3; Neurogenin-1; NGN-1
Gene Aliases: AKA; Ath4c; BHLHA6; Math4C; NEUROD3; NEUROG1; NGN; NGN1
UniProt ID: (Human) Q92886, (Mouse) P70660
Entrez Gene ID: (Human) 4762, (Mouse) 18014
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support