Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: NKKRMRGPGK DPTRKCPDPR SASSPKQHPS ESVYTALQRR ETEVYACIEN EDGSSPTAKQ SPLSQERPHR FEDDGELNLV YEN
The protein encoded by this gene is a type I membrane receptor that activates cytokine gene promoters such as the IL-13 and TNF-alpha promoters. The encoded protein contains an immunoreceptor tyrosine-based activation motif (ITAM) and is thought to regulate the signaling and development of B-cells. It is also known CNAIP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bK126B4.4; Calcineurin/NFAT-activating ITAM-containing protein; CTA-126B4.4; NFAT activation molecule 1; NFAT-activating protein with ITAM motif 1
Gene Aliases: CNAIP; NFAM1
UniProt ID: (Human) Q8NET5
Entrez Gene ID: (Human) 150372
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support