Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: A549, SiHa, A431.
Recognizes and binds the palindromic sequence 5'-TTGGCNNNNNGCCAA-3' present in viral and cellular promoters and in the origin of replication of adenovirus type 2. These proteins are individually capable of activating transcription and replication.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCAAT-box-binding transcription factor; CTF; KIAA1439; NF-I/C; NF1-C; NFI-A; NFI-B; NFI-C; NFI-X; NFIA; NFIB; NFIX; Nuclear factor 1 C-type; nuclear factor 1-C2; Nuclear factor 1/A; Nuclear factor 1/B; nuclear factor 1/C; Nuclear factor 1/X; Nuclear factor I/C; nuclear factor I/C (CCAAT-binding transcription factor); TGGCA-binding protein
Gene Aliases: 1110019L22Rik; 1500041O16Rik; AA589446; AI746521; CTF; CTF5; NF-I; NF1-C; NFI; NFIC
UniProt ID: (Human) P08651, (Mouse) P70255
Entrez Gene ID: (Human) 4782, (Mouse) 18029
Molecular Function: DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support