Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAGVACLGKA ADADEWCDSG LGSLGPDAAA PGGPGLGAEL GPGLSWAPLV FGYVTEDGDT ALHLAVIHQH EPFLDFLLGF SAGTEYMDLQ NDLGQTALHL AAILGETSTV EKLYAAGAGL CVAERRGHTA LHLACRVGAH ACARALLQPR PRRPREAPDT YLAQGPDRTP DTNHTPVALY PDSDLEKEEE ESEEDWKLQL EAENYEGHTP LHVAVIHKDV EMVRLLRDAG ADLDKPEPTC GRSPLHLAVE AQAADVLELL LRAGANPAAR MYGGRTPLGS AMLRPNPILA RLLRAHGAPE PEGEDEKSGP CSSSSDSDSG DEGDEYDDIV VHSSRSQTRL PPTPASKPLP DDPRPV
The NF-kB/Rel transcription factors are present in the cytosol in an inactive state, complexed with the inhibitory IkB proteins. Most agents that activate NF-kB do so through a common pathway based on phosphorylation-induced, proteasome-mediated degradation of IkB. The key regulatory step in this pathway involves activation of a high molecular weight IkB kinase (IKK) complex whose catalysis is generally carried out by three tightly associated IKK subunits. IKK alpha and IKK beta serve as the catalytic subunits of the kinase and IKK gamma serves as the regulatory subunit. Activation of IKK depends upon phosphorylation at Ser177 and Ser181 in the activation loop of IKK beta (Ser176 and Ser180 in IKK alpha), which causes conformational changes, resulting in kinase activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: I kappa B beta; I-kappa-B-beta; ikappaBbeta; IKB B; IKB beta; IkB-B; ikB-beta; IkBbeta; NF kappa BIB; NF-kappa-B inhibitor beta; NF-kappa-BIB; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta; Thyroid receptor; Thyroid receptor-interacting protein 9; TR-interacting protein 9; TRIP 9; TRIP-9
Gene Aliases: IKBB; NFKBIB; TRIP9
UniProt ID: (Human) Q15653
Entrez Gene ID: (Human) 4793
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support