Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MLLRAAWRR AAVAVTAAPG PKPAAPTRGL RLRVGDRAPQ SAVPADTAAA PEVGPVLRPL YMDVQATTPL DPRVLDAMLP YLINYYGNPH SRTHAYGWES EAAMERARQQ VASLIGADPR EIIFTSGATE SNNIAIKTES HSVIQAGVQW HDLDSLQAPP PGFTPFSCLS LLSSWDYRRP PPHPANFLYF (1-189 aa encoded by BC018471)
NFS1 (nitrogen fixation 1), also known as NIFS or IscS (cysteine desulfurase), is a member of the class V pyridoxal-phosphate-dependent aminotransferase family. It localizes to the cytoplasm or mitochondrion depending on which form is generated based on cytosolic pH. Highest expression levels of NFS1 are found in heart and skeletal muscle. Lower levels of expression are also found in liver, brain and pancreas. NFS1 is responsible for catalyzing the removal of sulfur from cysteine to form alanine, thereby supplying the inorganic sulfur for iron-sulfur (Fe-S) clusters. Fe-S clusters function as essential cofactors in a wide variety of events, including facilitation of electron transfer processes in oxidative phosphorylation, catalysis of enzymatic reactions in aconitase and dehydratases and maintenance of structural integrity in the DNA repair enzyme endonuclease III.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cysteine desulfurase; cysteine desulfurase, mitochondrial; m-Nfs1; NFS1 nitrogen fixation 1 homolog; nifS-like (sic); nitrogen fixation 1 (S. cerevisiae, homolog); nitrogen fixation gene 1; nitrogen fixation gene, yeast homolog 1; nitrogen-fixing bacteria S-like protein
Gene Aliases: AA987187; HUSSY-08; IscS; m-Nfs1; m-Nfsl; NFS1; NIFS
UniProt ID: (Human) Q9Y697, (Mouse) Q9Z1J3
Entrez Gene ID: (Human) 9054, (Rat) 84594, (Mouse) 18041
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support