Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: FKFNTDAAEF IPQEKKNSGL NCGTQRRLDS NRIGRRNYSS PPPCHLSRQV PYDEISAVHQ HSYHPSGSKP KSQQTSFQSS PCNKSPKSHG LQNQPWQKLR NEKHHIRVKK AQSLAEQTSD TAGLESSTRS ESGTDLREHS PSESEKEVVG ADPRGAKPKK ATQFVYSYGR GPKVKGKLKC EWSNRTTPKP EDAGPESTKP VGVFHPDSSE ASSRKGVLDG YGARRNEQRR YPQKRPPWEV EGARPRPGRN PPKQEGHRHT NAGHRNNMGP IPKDDLNERP AKSTCDSENL AVINKSSRRV DQEKCTVRRQ DPQVVSPFSR GKQNHVLKNV ETHTGSLIEQ
MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. NFX1 is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma.MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. The protein encoded by this gene is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma. Three alternative splice variants, each of which encodes a different isoform, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp779G2416; MGC20369; Nuclear transcription factor, X box-binding protein 1; Transcriptional repressor NF-X1; X-box binding 1
Gene Aliases: NFX1; NFX2; TEG-42; Tex42
UniProt ID: (Human) Q12986
Entrez Gene ID: (Human) 4799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support