FIGURE: 1 / 2
Involved in the translational repression of cytoplasmic polyadenylation element (CPE)-containing mRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CANu1; Centromere accumulated nuclear protein 1; EIF4E-binding protein; eukaryotic initiation factor 4E and CPEB binding protein; Neuroguidin; neuroguidin, EIF4E binding protein
Gene Aliases: C14orf120; CANu1; LCP5; lpd-2; NGD; NGDN
UniProt ID: (Human) Q8NEJ9
Entrez Gene ID: (Human) 25983
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support