Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Immunogen sequence: IRLMDIEDA KAHRRNKKDV NLSKTEKMGN TVESEHLSEL TEEEYEAHYI RRQDLKGFVW LDAKYLNPFF TRRLTQEDLH HGRIQMKTLT NKWYEEVRQG PSGSEDDEQE LL (471-581 aa encoded by BC112213)
Sodium-hydrogen exchangers (NHEs), such as SLC9A8, are integral transmembrane proteins that exchange extracellular Na+ for intracellular H+. NHEs have multiple functions, including intracellular pH homeostasis, cell volume regulation, and electroneutral NaCl absorption in epithelia (Xu et al., 2008).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686C03237; FLJ42500; MGC138418; Na(+)/H(+) exchanger 8; NHE-8; NHE-8 antibody; SLC9A8 antibody; Sodium/hydrogen exchanger 8; solute carrier family 9 (sodium/hydrogen exchanger), isoform 8; solute carrier family 9 (sodium/hydrogen exchanger), member 8; Solute carrier family 9 member 8; solute carrier family 9, subfamily A (NHE8, cation proton antiporter 8), member 8
Gene Aliases: 1200006P13Rik; 6430709P13Rik; AI182282; KIAA0939; NHE-8; NHE8; SLC9A8
UniProt ID: (Human) Q9Y2E8, (Rat) Q4L208, (Mouse) Q8R4D1
Entrez Gene ID: (Human) 23315, (Rat) 311651, (Mouse) 77031
Molecular Function:
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support