Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: MTKIKADPDG PEAQAEACSG ERTYQELLVN QNPIAQPLAS RRLTRKLYKC IKKAVKQKQI RRGVKEVQKF VNKGEKGIMV LAGDTLPIEV YCHLPVMCED RNLPYVYIPS KTDLGAAAGS KRPTCVIMVK PHEEYQEAYD ECLEEVQSLP LPL
NHP2 belongs to the H/ACA small nucleolar ribonucleoprotein (H/ACA snoRNP) complex, which catalyzes pseudouridylation of rRNA. It is required for ribosome biogenesis and telomere maintenance, and for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H/ACA ribonucleoprotein complex subunit 2; NHP2 ribonucleoprotein; NHP2 ribonucleoprotein homolog; NHP2-like protein; Nucleolar protein family A member 2; nucleolar protein family A, member 2; nucleolar protein family A, member 2 (H/ACA small nucleolar RNPs); snoRNP protein NHP2
Gene Aliases: DKCB2; HSPC286; NHP2; NHP2P; NOLA2
UniProt ID: (Human) Q9NX24
Entrez Gene ID: (Human) 55651, (Rat) 287273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support