Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: RELSAPARLY HKKVVDHYEN PRNVGSLDKT SKNVGTGLVG APACGDVMKL QIQVDEKGKI VDARFKTFGC GSAIASSSLA TEWVKGKTVE EALTIKNTDI AKELCLPPVK LHCSMLAEDA IKAALADYKL KQEPKKGEAE KK
Iron-sulfur (Fe-S) clusters are necessary for several mitochondrial enzymes and other subcellular compartment proteins. They contain sulfur and iron, and are created via several steps that include cysteine desulfurases, iron donors, chaperones, and scaffold proteins. This gene encodes the two isomeric forms, ISCU1 and ISCU2, of the Fe-S cluster scaffold protein. Mutations in this gene have been found in patients with myopathy with severe exercise intolerance and myoglobinuria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Iron-sulfur cluster assembly enzyme ISCU; IscU iron-sulfur cluster scaffold homolog; NifU-like N-terminal domain-containing protein; NifU-like protein
Gene Aliases: 2310020H20Rik; HML; hnifU; ISCU; ISU2; NIFU; NIFUN
UniProt ID: (Human) Q9H1K1
Entrez Gene ID: (Human) 23479
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support