Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Pig and Orangutan
NIP7 contains 1PUA domain and belongs to the NIP7 family. It may play a role in 60S ribosomal subunit synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosome subunit biogenesis protein NIP7 homolog; FLJ10296; KD93; kDa93; nuclear import 7 homolog; Nucleolar pre-rRNA processing protein NIP7; PEachy
Gene Aliases: 1110017C15Rik; 6330509M23Rik; AA408773; AA410017; CGI-37; HSPC031; HSPC180; KD93; NIP7; OK/SW-cl.76; OK/SW-cl.78; PEachy
UniProt ID: (Human) Q9Y221, (Mouse) Q9CXK8
Entrez Gene ID: (Human) 51388, (Mouse) 66164
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support